2-(2-Thienyl)-4-thiazolecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Thienyl)-1,3-thiazole-4-carboxylicacid

044083-500MG 044083-1G 044083-5G
101864-042EA 113.85 USD
101864-042 101864-044 101864-046
2-(2-Thienyl)-4-thiazolecarboxylic acid ≥95%
2-(2-Thienyl)-4-thiazolecarboxylic acid

Hazard: Irritant

Formula: C₈H₅NO₂S₂
MW: 211.25 g/mol
Melting Pt: 165-167°C
MDL Number: MFCD01936014
CAS Number: 24044-07-3

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR