Ethyl-2-(2-thienyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(thiophen-2-yl)thiazole-4-carboxylate

044082-500MG 044082-1G 044082-5G
101864-036EA 556.92 USD
101864-036 101864-038 101864-040
Ethyl-2-(2-thienyl)-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-(2-thienyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₂S₂
MW: 239.32 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD00107415
CAS Number: 24043-97-8

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR