Ethyl-2-[2-(aminocarbonyl)hydrazono]-3-[(4-chlorophenyl)sulfanyl]propanoate ≥95%

Supplier: Matrix Scientific

044081-500MG 044081-1G
101864-032EA 278.46 USD
101864-032 101864-034
Ethyl-2-[2-(aminocarbonyl)hydrazono]-3-[(4-chlorophenyl)sulfanyl]propanoate ≥95%
Ethyl-2-[2-(aminocarbonyl)hydrazono]-3-[(4-chlorophenyl)sulfanyl]propanoate

Hazard: Irritant

Formula: C₁₂H₁₄ClN₃O₃S
Melting Pt: 153…156 °C
MDL Number: MFCD08056619
CAS Number: 886361-57-5

Order Now

Specification Test Results

Melting point 153-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR