2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

044078-500MG 044078-1G 044078-5G
101864-014EA 375.38 USD
101864-014 101864-016 101864-018
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitrile ≥95%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(4-methoxyphenyl)acetonitrile

Hazard: Irritant

Formula: C₁₅H₁₀ClF₃N₂O
Melting Pt: 100-102°C
MDL Number: MFCD00172212
CAS Number: 338407-10-6

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR