2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(2,6-dichlorophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

044076-500MG 044076-1G 044076-5G
101864-002EA 375.38 USD
101864-002 101864-004 101864-006
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(2,6-dichlorophenyl)acetonitrile ≥95%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-(2,6-dichlorophenyl)acetonitrile

Hazard: Irritant

Formula: C₁₄H₆Cl₃F₃N₂
Melting Pt: 107…109 °C
MDL Number: MFCD00172210
CAS Number: 338407-06-0

Order Now

Specification Test Results

Melting point 107-109°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR