2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile ≥95%
Supplier: Matrix Scientific
044075-500MG
044075-1G
044075-5G
101863-996EA
307.69
USD
101863-996
101863-998
101864-000
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile ≥95%
2-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]-2-phenylacetonitrile
Hazard: Irritant
Formula:
C₁₄H₈ClF₃N₂ Melting Pt: 70…72 °C |
MDL Number:
MFCD00172206 CAS Number: 213994-27-5 |
Specification Test Results
Melting point | 70-72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...