1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride ≥95%
Supplier: Matrix Scientific
Synonyms:
O-(4-Fluorobenzyl)hydroxylamine hydrochloride
044072-1G
044072-5G
101863-986EA
275.38
USD
101863-986
101863-988
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-4-fluorobenzene chloride
Hazard: Irritant
Formula:
C₇H₉ClFNO MW: 177.6039 g/mol Melting Pt: 225-235°C |
MDL Number:
MFCD01114581 CAS Number: 51572-89-5 |
Specification Test Results
Melting point | 225-235°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...