1-(2-(Ammoniooxy)ethoxy)benzene chloride ≥95%

Supplier: Matrix Scientific

044071-500MG 044071-1G 044071-5G
101863-980EA 375.38 USD
101863-980 101863-982 101863-984
1-(2-(Ammoniooxy)ethoxy)benzene chloride ≥95%
1-(2-(Ammoniooxy)ethoxy)benzene chloride

Hazard: Irritant

Formula: C₈H₁₂ClNO₂
Melting Pt: 182-188°C
MDL Number: MFCD01114593
CAS Number: 5397-72-8

Order Now

Specification Test Results

Melting point 182-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR