Ethyl-2-amino-4-(4-chlorophenyl)-1H-pyrrole-3-carboxylate ≥95%
Supplier: Matrix Scientific
044069-500MG
044069-1G
101863-972EA
1483.08
USD
101863-972
101863-974
Ethyl-2-amino-4-(4-chlorophenyl)-1H-pyrrole-3-carboxylate ≥95%
Ethyl-2-amino-4-(4-chlorophenyl)-1H-pyrrole-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₃ClN₂O₂ Melting Pt: 144…146 °C |
MDL Number:
MFCD00244763 CAS Number: 309713-15-3 |
Specification Test Results
Melting point | 144-146°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...