1H-Pyrrole-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

044062-500MG 044062-1G 044062-5G
101863-938EA 283.08 USD
101863-938 101863-940 101863-942
1H-Pyrrole-2-carbohydrazide ≥95%
1H-Pyrrole-2-carbohydrazide

Hazard: Irritant

Formula: C₅H₇N₃O
Melting Pt: 236…238 °C
MDL Number: MFCD00141885
CAS Number: 50269-95-9

Order Now

Specification Test Results

Melting point 236-238°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR