2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: Oxalic monohydrazide mono(4-trifluoromethoxy)anilide

044060-500MG 044060-1G 044060-5G
101863-926EA 375.38 USD
101863-926 101863-928 101863-930
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide ≥95%
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide

Hazard: Irritant

Formula: C₉H₈F₃N₃O₃
MW: 263.18 g/mol
Melting Pt: 215…217 °C
MDL Number: MFCD00173093
CAS Number: 338405-24-6

Order Now

Specification Test Results

Melting point 215-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR