Methyl-6-oxo-4-phenoxy-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

044059-500MG 044059-1G 044059-5G
101863-920EA 556.92 USD
101863-920 101863-922 101863-924
Methyl-6-oxo-4-phenoxy-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-6-oxo-4-phenoxy-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₈H₁₄N₂O₄
Melting Pt: 128…130 °C
MDL Number: MFCD00141171
CAS Number: 338405-06-4

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR