2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid ≥95%

Supplier: Matrix Scientific

044058-1G 044058-5G
101863-916EA 113.85 USD
101863-916 101863-918
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid ≥95%
2-{[3-(Trifluoromethyl)benzoyl]amino}acetic acid

Hazard: Irritant

Formula: C₁₀H₈F₃NO₃
Melting Pt: 134…138 °C
MDL Number: MFCD00029782
CAS Number: 17794-48-8

Order Now

Specification Test Results

Melting point 134-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR