2-[(4-Methoxybenzoyl)amino]acetic acid ≥95%
Supplier: Matrix Scientific
044057-500MG
044057-1G
044057-5G
101863-910EA
109.23
USD
101863-910
101863-912
101863-914
2-[(4-Methoxybenzoyl)amino]acetic acid ≥95%
2-[(4-Methoxybenzoyl)amino]acetic acid
Hazard: Irritant
Formula:
C₁₀H₁₁NO₄ Melting Pt: 155…158 °C |
MDL Number:
MFCD00181815 CAS Number: 13214-64-7 |
Specification Test Results
Melting point | 155-158°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...