Methyl-3-(benzoylamino)-6-methyl-2-oxo-2H-pyran-5-carboxylate ≥95%

Supplier: Matrix Scientific

044055-500MG 044055-1G 044055-5G
101863-898EA 278.46 USD
101863-898 101863-900 101863-902
Methyl-3-(benzoylamino)-6-methyl-2-oxo-2H-pyran-5-carboxylate ≥95%
Methyl-3-(benzoylamino)-6-methyl-2-oxo-2H-pyran-5-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₃NO₅
Melting Pt: 170…173 °C
MDL Number: MFCD00139779
CAS Number: 338404-89-0

Order Now

Specification Test Results

Melting point 170-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR