4-Chloro-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

044052-500MG 044052-1G 044052-5G
101863-880EA 375.38 USD
101863-880 101863-882 101863-884
4-Chloro-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-2-(2-pyridinyl)-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₅ClF₃N₃
MW: 259.62 g/mol
Melting Pt: 46…47 °C
MDL Number: MFCD11100198
CAS Number: 438249-84-4

Order Now

Specification Test Results

Melting point 46-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR