2-Methyl-2-(4-phenylpiperazino)propanenitrile ≥95%

Supplier: Matrix Scientific

044049-500MG 044049-1G 044049-5G
101863-862EA 181.54 USD
101863-862 101863-864 101863-866
2-Methyl-2-(4-phenylpiperazino)propanenitrile ≥95%
2-Methyl-2-(4-phenylpiperazino)propanenitrile

Hazard: Irritant

Formula: C₁₄H₁₉N₃
MW: 229.33 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00202004
CAS Number: 92326-91-5

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR