4-(2-Chlorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

044040-500MG 044040-1G 044040-5G
101863-816EA 375.38 USD
101863-816 101863-818 101863-820
4-(2-Chlorophenoxy)-3-nitrobenzenecarbaldehyde ≥95%
4-(2-Chlorophenoxy)-3-nitrobenzenecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClNO₄
Melting Pt: 81…84 °C
MDL Number: MFCD00138771
CAS Number: 338403-42-2

Order Now

Specification Test Results

Melting point 81-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR