1-(4-Chlorophenyl)-2-[(4-chlorophenyl)sulfanyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
044039-500MG
044039-1G
044039-5G
101863-810EA
181.54
USD
101863-810
101863-812
101863-814
1-(4-Chlorophenyl)-2-[(4-chlorophenyl)sulfanyl]-1-ethanone ≥95%
1-(4-Chlorophenyl)-2-[(4-chlorophenyl)sulfanyl]-1-ethanone
Hazard: Irritant
Formula:
C₁₄H₁₀Cl₂OS Melting Pt: 112…115 °C |
MDL Number:
MFCD00098687 CAS Number: 33046-50-3 |
Specification Test Results
Melting point | 112-115°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...