Ethyl-4-amino-3-methylthieno[2,3-c]isothiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
044038-500MG
044038-1G
044038-5G
101863-804EA
375.38
USD
101863-804
101863-806
101863-808
Ethyl-4-amino-3-methylthieno[2,3-c]isothiazole-5-carboxylate ≥95%
Ethyl-4-amino-3-methylthieno[2,3-c]isothiazole-5-carboxylate
Hazard: Irritant
Formula:
C₉H₁₀N₂O₂S₂ Melting Pt: 134…136 °C |
MDL Number:
MFCD00139775 CAS Number: 82000-54-2 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...