7-Amino-2-(methylsulfanyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

044037-500MG 044037-1G 044037-5G
101863-798EA 747.69 USD
101863-798 101863-800 101863-802
7-Amino-2-(methylsulfanyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%
7-Amino-2-(methylsulfanyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylic acid

Hazard: Irritant

Formula: C₇H₇N₅O₂S
MW: 225.23 g/mol
Melting Pt: 275 °C
MDL Number: MFCD00067817
CAS Number: 113967-71-8

Order Now

Specification Test Results

Melting point 275°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR