4-[(1,3,5-Trimethyl-1H-pyrazol-4-yl)methyl]benzenol ≥95%

Supplier: Matrix Scientific

044036-500MG 044036-1G 044036-5G
101863-792EA 181.54 USD
101863-792 101863-794 101863-796
4-[(1,3,5-Trimethyl-1H-pyrazol-4-yl)methyl]benzenol ≥95%
4-[(1,3,5-Trimethyl-1H-pyrazol-4-yl)methyl]benzenol

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MW: 216.29 g/mol
Melting Pt: 170…171 °C
MDL Number: MFCD00138770
CAS Number: 75999-00-7

Order Now

Specification Test Results

Melting point 170-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR