2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: (2Z)-2-(4-Chlorophenyl)-3-(dimethylamino)prop-2-enenitrile

044035-500MG 044035-1G 044035-5G
101863-786EA 181.54 USD
101863-786 101863-788 101863-790
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(4-Chlorophenyl)-3-(dimethylamino)acrylonitrile

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂
MW: 206.68 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD00138415
CAS Number: 40373-88-4

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR