2-(Methylsulfinyl)phenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxyphenyl methyl sulphoxide

044033-500MG 044033-1G
101863-776EA 138.46 USD
101863-776 101863-778
2-(Methylsulfinyl)phenol ≥95%
2-(Methylsulphinyl)phenol

Hazard: Irritant

Formula: C₇H₈O₂S
Melting Pt: 126-128°C
MDL Number: MFCD00127350
CAS Number: 1074-02-8

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR