Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate ≥95%

Supplier: Matrix Scientific

044030-500MG 044030-1G
101863-764EA 432.31 USD
101863-764 101863-766
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate ≥95%
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetate

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂
MW: 253.61 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD07366545
CAS Number: 885949-63-3

Order Now

Specification Test Results

Melting point 38-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR