5-Amino-3-(4-bromophenyl)isoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)isoxazol-5-amine

044029-500MG 044029-1G 044029-5G
101863-758EA 190.77 USD
101863-758 101863-760 101863-762
5-Amino-3-(4-bromophenyl)isoxazole ≥95%
5-Amino-3-(4-bromophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₇BrN₂O
MW: 239.700 g/mol
Melting Pt: 160-163°C
MDL Number: MFCD06199354
CAS Number: 119162-53-7

Order Now

Specification Test Results

Melting point 160-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR