5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole ≥95%
Supplier: Matrix Scientific
044027-500MG
044027-1G
044027-5G
101863-746EA
375.38
USD
101863-746
101863-748
101863-750
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole ≥95%
5-Bromo-3-(1,2,3,6-tetrahydro-4-pyridinyl)-1H-indole
Hazard: Irritant
Formula:
C₁₃H₁₃BrN₂ MW: 277.17 g/mol Melting Pt: 190…192 °C |
MDL Number:
MFCD07366543 CAS Number: 127792-80-7 |
Specification Test Results
Melting point | 190-192°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...