N-{4-[(2-Cyanobenzyl)oxy]phenyl}acetamide ≥95%

Supplier: Matrix Scientific

044025-500MG 044025-1G 044025-5G
101863-734EA 181.54 USD
101863-734 101863-736 101863-738
N-{4-[(2-Cyanobenzyl)oxy]phenyl}acetamide ≥95%
N-{4-[(2-Cyanobenzyl)oxy]phenyl}acetamide

Hazard: Irritant

Formula: C₁₆H₁₄N₂O₂
Melting Pt: 144…146 °C
MDL Number: MFCD06672957
CAS Number: 852386-93-7

Order Now

Specification Test Results

Melting point 144-146°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR