6-Chloro-N-(2-hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide ≥95%

Supplier: Matrix Scientific

044024-500MG 044024-1G
101863-730EA 556.92 USD
101863-730 101863-732
6-Chloro-N-(2-hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide ≥95%
6-Chloro-N-(2-hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide

Hazard: Irritant

Formula: C₁₀H₁₀ClN₃O₂
Melting Pt: 176…178 °C
MDL Number: MFCD07366542
CAS Number: 885949-61-1

Order Now

Specification Test Results

Melting point 176-178°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR