Ethyl-2-aminothiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-aminothiazole-5-carboxylate

044023-5G 044023-25G
101863-726EA 21.54 USD
101863-726 101863-728
Ethyl-2-aminothiazole-5-carboxylate ≥95%
Ethyl-2-aminothiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₈N₂O₂S
Melting Pt: 158…160 °C
MDL Number: MFCD00602139
CAS Number: 32955-21-8

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR