5-(Phenylsulfanyl)-2-furaldehyde ≥95%
Supplier: Matrix Scientific
044022-500MG
044022-1G
101863-720EA
375.38
USD
101863-720
101863-722
5-(Phenylsulfanyl)-2-furaldehyde ≥95%
5-(Phenylsulfanyl)-2-furaldehyde
Hazard: Irritant
Formula:
C₁₁H₈O₂S Melting Pt: 39…40 °C |
MDL Number:
MFCD02629617 CAS Number: 39689-03-7 |
Specification Test Results
Melting point | 39-40°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...