6-Oxo-1,6-dihydropyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

044021-500MG 044021-1G
101863-716EA 146.15 USD
101863-716 101863-718
6-Oxo-1,6-dihydropyridine-3-carbonitrile ≥95%
6-Hydroxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₆H₄N₂O
MW: 120.109 g/mol
Melting Pt: 255…257 °C
MDL Number: MFCD07698589
CAS Number: 94805-52-4

Order Now

Specification Test Results

Melting point 255-257°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR