1-(3,4-Dichlorophenyl)-2-hydroxy-1-ethanone ≥95%

Supplier: Matrix Scientific

044016-500MG 044016-1G 044016-5G
101863-688EA 375.38 USD
101863-688 101863-690 101863-692
1-(3,4-Dichlorophenyl)-2-hydroxy-1-ethanone ≥95%
1-(3,4-Dichlorophenyl)-2-hydroxy-1-ethanone

Hazard: Irritant

Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD07698585
CAS Number: 113337-38-5

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR