5-[(4-Bromophenyl)sulfanyl]-2-furaldehyde ≥95%

Supplier: Matrix Scientific

044015-500MG 044015-1G 044015-5G
101863-682EA 278.46 USD
101863-682 101863-684 101863-686
5-[(4-Bromophenyl)sulfanyl]-2-furaldehyde ≥95%
5-[(4-Bromophenyl)sulfanyl]-2-furaldehyde

Hazard: Irritant

Formula: C₁₁H₇BrO₂S
Melting Pt: 78…80 °C
MDL Number: MFCD07698584
CAS Number: 56656-94-1

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR