2-Cyano-3-(3-pyridinyl)acrylic acid ≥95%
Supplier: Matrix Scientific
044008-500MG
044008-1G
044008-5G
101863-648EA
181.54
USD
101863-648
101863-650
101863-652
2-Cyano-3-(3-pyridinyl)acrylic acid ≥95%
2-Cyano-3-(3-pyridinyl)acrylic acid
Hazard: Irritant
Formula:
C₉H₆N₂O₂ Melting Pt: 195 °C |
MDL Number:
MFCD02668152 CAS Number: 103029-74-9 |
Specification Test Results
Melting point | 195°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...