2-Cyano-3-(2-thienyl)acrylic acid ≥95%

Supplier: Matrix Scientific

044007-500MG 044007-1G 044007-5G
101863-642EA 147.69 USD
101863-642 101863-644 101863-646
2-Cyano-3-(2-thienyl)acrylic acid ≥95%
2-Cyano-3-(2-thienyl)acrylic acid

Hazard: Irritant

Formula: C₈H₅NO₂S
MW: 179.2 g/mol
Melting Pt: 217…220 °C
MDL Number: MFCD03412481
CAS Number: 58177-53-0

Order Now

Specification Test Results

Melting point 217-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR