Ethyl-2-cyano-3-(4-methylphenyl)acrylate ≥95%
Supplier: Matrix Scientific
044006-1G
044006-500MG
044006-5G
101863-638EA
232.31
USD
101863-638
101863-640
101863-636
Ethyl-2-cyano-3-(4-methylphenyl)acrylate ≥95%
Ethyl-2-cyano-3-(4-methylphenyl)acrylate
Hazard: Irritant
Formula:
C₁₃H₁₃NO₂ MW: 215.25 g/mol Melting Pt: 86…89 °C |
MDL Number:
MFCD00136329 CAS Number: 2017-88-1 |
Specification Test Results
Melting point | 86-89°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...