Ethyl-2-cyano-3-(2-thienyl)acrylate ≥95%

Supplier: Matrix Scientific

044004-500MG 044004-1G 044004-5G
101863-630EA 181.54 USD
101863-630 101863-632 101863-634
Ethyl-2-cyano-3-(2-thienyl)acrylate ≥95%
Ethyl-2-cyano-3-(2-thienyl)acrylate

Hazard: Irritant

Formula: C₁₀H₉NO₂S
MW: 207.25 g/mol
Melting Pt: 79…82 °C
MDL Number: MFCD00039684
CAS Number: 31330-51-5

Order Now

Specification Test Results

Melting point 79-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR