3-(1,3-Benzodioxol-5-yl)-2-cyanoacrylic acid ≥95%

Supplier: Matrix Scientific

044003-500MG 044003-1G 044003-5G
101863-624EA 181.54 USD
101863-624 101863-626 101863-628
3-(1,3-Benzodioxol-5-yl)-2-cyanoacrylic acid ≥95%
3-(1,3-Benzodioxol-5-yl)-2-cyanoacrylic acid

Hazard: Irritant

Formula: C₁₁H₇NO₄
MW: 217.18 g/mol
Melting Pt: 170 °C
MDL Number: MFCD00759392
CAS Number: 49711-55-9

Order Now

Specification Test Results

Melting point 170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR