Ethyl-3-(1,3-benzodioxol-5-yl)-2-cyanoacrylate ≥95%

Supplier: Matrix Scientific

044002-500MG 044002-1G 044002-5G
101863-618EA 181.54 USD
101863-618 101863-620 101863-622
Ethyl-3-(1,3-benzodioxol-5-yl)-2-cyanoacrylate ≥95%
Ethyl-3-(1,3-benzodioxol-5-yl)-2-cyanoacrylate

Hazard: Irritant

Formula: C₁₃H₁₁NO₄
Melting Pt: 95…97 °C
MDL Number: MFCD00022988
CAS Number: 2286-56-8

Order Now

Specification Test Results

Melting point 95-97°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR