Ethyl 2-cyano-3-(pyridin-3-yl)acrylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl-2-cyano-3-(3-pyridinyl)acrylate

044001-500MG 044001-1G 044001-5G
101863-612EA 181.54 USD
101863-612 101863-614 101863-616
Ethyl 2-cyano-3-(pyridin-3-yl)acrylate ≥95%
Ethyl 2-cyano-3-(pyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.209 g/mol
Melting Pt: 67…70 °C
MDL Number: MFCD00564622
CAS Number: 17999-71-2

Order Now

Specification Test Results

Melting point 67-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR