6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid ≥95%
Supplier: Matrix Scientific
044000-500MG
044000-1G
044000-5G
101863-606EA
181.54
USD
101863-606
101863-608
101863-610
6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid ≥95%
6-Chloro-2-(4-hydroxyphenyl)-3-methyl-4-quinolinecarboxylic acid
Hazard: Irritant
Formula:
C₁₇H₁₂ClNO₃ MW: 313.74 g/mol Melting Pt: 350 °C |
MDL Number:
MFCD06200950 CAS Number: 854867-53-1 |
Specification Test Results
Melting point | 350°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...