Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

043996-500MG 043996-1G 043996-5G
101863-588EA 278.46 USD
101863-588 101863-590 101863-592
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate ≥95%
Methyl-1-(4,6-dimethyl-2-pyrimidinyl)-1H-1,2,3-triazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁N₅O₂
MW: 233.23 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00838109
CAS Number: 23947-13-9

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR