[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
4,5-Bis(hydroxymethyl)-1-(4,6-dimethylpyrimidin-2-yl)-1H-1,2,3-triazole
043995-500MG
043995-1G
043995-5G
101863-582EA
278.46
USD
101863-582
101863-584
101863-586
[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol ≥95%
[1-(4,6-Dimethyl-2-pyrimidinyl)-5-(hydroxymethyl)-1H-1,2,3-triazol-4-yl]methanol
Hazard: Irritant
Formula:
C₁₀H₁₃N₅O₂ MW: 235.25 g/mol Melting Pt: 172…174 °C |
MDL Number:
MFCD06200948 CAS Number: 886361-66-6 |
Specification Test Results
Melting point | 172-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...