1-(3-Ethynylphenyl)-3,3-dimethyl-2-azetanone ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3-Dimethyl-1-(3-ethynylphenyl)azetidin-2-one

043992-500MG 043992-1G 043992-5G
101863-564EA 347.69 USD
101863-564 101863-566 101863-568
1-(3-Ethynylphenyl)-3,3-dimethyl-2-azetanone ≥95%
1-(3-Ethynylphenyl)-3,3-dimethyl-2-azetanone

Hazard: Irritant

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD06200947
CAS Number: 886361-64-4

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR