Ethyl-5-bromo-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

043988-500MG 043988-1G 043988-5G
101863-542EA 190.77 USD
101863-542 101863-544 101863-546
Ethyl-5-bromo-1-benzofuran-2-carboxylate ≥95%
Ethyl-5-bromo-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₉BrO₃
Melting Pt: 60…62 °C
MDL Number: MFCD02667598
CAS Number: 84102-69-2

Order Now

Specification Test Results

Melting point 60-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR