1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Bromoacetyl)benzo[b]thiophene

043987-500MG 043987-1G 043987-5G
101863-536EA 556.92 USD
101863-536 101863-538 101863-540
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone ≥95%
1-(1-Benzothiophen-3-yl)-2-bromo-1-ethanone

Hazard: Irritant

Formula: C₁₀H₇BrOS
Melting Pt: 142…144 °C
MDL Number: MFCD00176410
CAS Number: 26167-45-3

Order Now

Specification Test Results

Melting point 142-144°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR