1-[(6-Chloro-3-pyridinyl)methyl]-4-(2,6-dimethylphenyl)piperazine ≥95%

Supplier: Matrix Scientific

043980-500MG 043980-1G 043980-5G
101863-500EA 278.46 USD
101863-500 101863-502 101863-504
1-[(6-Chloro-3-pyridinyl)methyl]-4-(2,6-dimethylphenyl)piperazine ≥95%
1-[(6-Chloro-3-pyridinyl)methyl]-4-(2,6-dimethylphenyl)piperazine

Hazard: Irritant

Formula: C₁₈H₂₂ClN₃
Melting Pt: 100…102 °C
MDL Number: MFCD04125008
CAS Number: 861211-43-0

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR