Methyl-3-(4-aminophenoxy)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
043975-500MG
043975-1G
043975-5G
101863-482EA
556.92
USD
101863-482
101863-484
101863-486
Methyl-3-(4-aminophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(4-aminophenoxy)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁NO₃S Melting Pt: 102…105 °C |
MDL Number:
MFCD04117804 CAS Number: 103790-38-1 |
Specification Test Results
Melting point | 102-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...