Methyl-3-(2,4-diaminophenoxy)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043974-500MG 043974-1G 043974-5G
101863-476EA 556.92 USD
101863-476 101863-478 101863-480
Methyl-3-(2,4-diaminophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(2,4-diaminophenoxy)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₃S
Melting Pt: 166…169 °C
MDL Number: MFCD04117803
CAS Number: 104636-77-3

Order Now

Specification Test Results

Melting point 166-169°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR